Flavin adenine dinucleotide disodium salt

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

347212500 347210010
ACRO347212500EA 126 EUR
ACRO347212500 ACRO347210010
Flavin adenine dinucleotide disodium salt
Flavin adenine dinucleotide disodium salt

CAS No.: 84366-81-4

Formule: C₂₇H₃₃N₉O₁₅P₂
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00151217
CAS nummer: 84366-81-4

Order Now

Specification Test Results

Appearance Yellow to orange Powder
Infrared spectrum Conforms
Water ≤10 % (K.F.)
UV spectrophotometry molar absorptivity 11300 +/- 4%, on dry substance


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR