tert-Butyl chloroacetate 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

347230250 347231000 347235000
ACRO347230250EA 25 EUR
ACRO347230250 ACRO347231000 ACRO347235000
tert-Butyl chloroacetate 97%
tert-Butyl chloroacetate
Formule: C₆H₁₁ClO₂
Kookpunt: 48…49 °C (11 mmHg)
Dichtheid: 1,053 g/cm³
Vlampunt: 46 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000930
CAS nummer: 107-59-5
EINECS: 203-506-1
UN: 2929
ADR: 6.1,II
Merck Index: 13,01562

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4220 to 1.4240 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR