Succinimide 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

348650050 348651000 348655000
ACRO348650050EA 29.7 EUR
ACRO348650050 ACRO348651000 ACRO348655000
Succinimide 98%
Succinimide
Formule: C₄H₅NO₂
Kookpunt: 285…290 °C (760 mmHg)
Smeltpunt: 121…126 °C
Dichtheid: 1,418 g/cm³ (20 °C)
Vlampunt: 201 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005495
CAS nummer: 123-56-8
EINECS: 204-635-6
Merck Index: 12,09040

Order Now

Specification Test Results

Appearance Off-white to beige to light brown Powder or flakes
Infrared spectrum Conforms
Melting point 121 to 126 °C
Titration with NaOH ≥97.5 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR