1,1-Di-(tert-butylperoxy)-3,3,5-trimethylcyclohexane 75%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

349772500
ACRO349772500EA 88.2 EUR
ACRO349772500
1,1-Di-(tert-butylperoxy)-3,3,5-trimethylcyclohexane 75%
1,1-Di-(tert-butylperoxy)-3,3,5-trimethylcyclohexane

Appearance: Clear solution

Formule: C₁₇H₃₄O₄
Kookpunt: –20 °C
Smeltpunt: –20 °C
Dichtheid: 0,87 g/cm³
Vlampunt: 62 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00084801
CAS nummer: 6731-36-8
UN: 3103
ADR: 5.2,

Order Now

Specification Test Results

Appearance Clear solution
Titration Iodometric 74 to 76 %
Color scale ≤50 APHA
Impurity ≤0.8 % Hydroperoxides (as TBHP)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR