4-Methoxybenzaldehyde dimethyl acetal 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: p-(Dimethoxymethyl)anisole, P-anisaldehyde dimethyl acetal, p-Anisaldehyde dimethyl acetal, 1-(Dimethoxymethyl)-4-methoxybenzene, 4-Methoxy-α,α-dimethoxytoluene

352290250 352291000 352295000
ACRO352290250EA 52.5 EUR
ACRO352290250 ACRO352291000 ACRO352295000
4-Methoxybenzaldehyde dimethyl acetal 98%
4-Methoxybenzaldehyde dimethyl acetal
Formule: C₁₀H₁₄O₃
MW: 182.22 g/mol
Kookpunt: 235 °C (760 mmHg)
Dichtheid: 1.068 g/cm³
Vlampunt: >110 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00036507
CAS nummer: 2186-92-7
EINECS: 218-577-4

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5035 to 1.5075 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR