Pyridoxal hydrochloride 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

352710050 352710010 352710250 352711000 352715000
ACRO352710050EA 74 EUR
ACRO352710050 ACRO352710010 ACRO352710250 ACRO352711000 ACRO352715000
Pyridoxal hydrochloride 99%
Pyridoxal hydrochloride
Formule: C₈H₁₀ClNO₃
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00012809
CAS nummer: 65-22-5
EINECS: 200-602-5

Order Now

Specification Test Results

Appearance White to yellow to light beige Powder
Infrared spectrum Conforms
Melting point 171 to 175 °C (Dec)
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR