6-Bromopiperonal 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 6-Bromobenzo[d][1,3]dioxole-5-carbaldehyde

355920010 355920050 355920250
ACRO355920010EA 29.7 EUR
ACRO355920010 ACRO355920050 ACRO355920250
6-Bromopiperonal 98%
6-Bromopiperonal
Formule: C₈H₅BrO₃
MW: 229.03 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00022952
CAS nummer: 15930-53-7

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 125 to 129 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR