(R)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride 95%, 98% ee

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

362990010 362990050
ACRO362990010EA 191 EUR
ACRO362990010 ACRO362990050
(R)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride 95%, 98% ee
(R)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride

Appearance: White Powder

Formule: C₁₁H₁₆ClNO₂
Smeltpunt: 119 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD02259750
CAS nummer: 340188-50-3

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
HPLC ≥94 %
Enantiomeric excess ≥97.5 % (GC)

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR