Dichloro(p-cymene)ruthenium(II) dimer 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Di(μ-chloro)dichlorobis(p-cymene)diruthenium(II)

363290010 363290050
ACRO363290010EA 65.1 EUR
ACRO363290010 ACRO363290050
Dichloro(p-cymene)ruthenium(II) dimer 98%
Dichloro(p-cymene)ruthenium(II) dimer

CAS No.: 52462-29-0

Formule: C₂₀H₂₈Cl₄Ru₂
MW: 612.38 g/mol
Smeltpunt: 250 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064793
CAS nummer: 52462-29-0

Order Now

Specification Test Results

Appearance Orange-red to red-brown Powder or crystals
Infrared spectrum Conforms
Elemental Analysis ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR