Zinc fluoride 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

363910250 363912500
ACRO363910250EA 52 EUR
ACRO363910250 ACRO363912500
Zinc fluoride 98%
Zinc fluoride
Formule: F₂Zn
MW: 103.39 g/mol
Kookpunt: 1500
Smeltpunt: 872…910 °C
Dichtheid: 4.95
MDL Nummer: MFCD00011298
CAS nummer: 7783-49-5
EINECS: 232-001-9

Order Now

Specification Test Results

Appearance White to off-white Powder
Fluoride (F) 36.0 to 37.8 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR