tert-Butyl-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate 97+%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

366350010 366350050
ACRO366350010EA 32 EUR
ACRO366350010 ACRO366350050
tert-Butyl-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate 97+%
tert-Butyl-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate

Appearance: White to off-white Crystalline powder

Formule: C₁₇H₂₆BNO₄
Smeltpunt: 166…169 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD02179439
CAS nummer: 330793-01-6

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 168 to 173 °C
HPLC ≥97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR