Zoutzuur 4 N in 1,4-dioxane/water

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

366521000 366520025
ACRO366521000EA 72.7 EUR
ACRO366521000 ACRO366520025
Zoutzuur 4 N in 1,4-dioxane/water
Zoutzuur
Formule: HCl
Smeltpunt: -30 °C
Dichtheid: 1,1 g/cm³
Vlampunt: 12 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00011324
CAS nummer: 7647-01-0
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Solution
Strength 3.95 to 4.40 N


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR