Zink 99.999% (trace metals basis), powder 40 mesh

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

367260100 367260500
ACRO367260100EA 198 EUR
ACRO367260100 ACRO367260500
Zink 99.999% (trace metals basis), powder 40 mesh
Zink
Formule: Zn
Kookpunt: 907 °C (1013 hPa)
Smeltpunt: 420 °C
Dichtheid: 7,13 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011291
CAS nummer: 7440-66-6
EINECS: 231-175-3
UN: 1436
ADR: 4.3,II

Order Now

Specification Test Results

Appearance Grey Powder
Total trace metal impurities ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR