(Trifluoromethyl)trimethylsilane 0.5 M in THF

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

368320050 368320250 368321000
ACRO368320050EA 21.8 EUR
ACRO368320050 ACRO368320250 ACRO368321000
(Trifluoromethyl)trimethylsilane 0.5 M in THF
(Trifluoromethyl)trimethylsilane
Formule: C₄H₉F₃Si
Kookpunt: 40 °C (760 mmHg)
Dichtheid: 0,895 g/cm³
Vlampunt: –20 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00145454
CAS nummer: 81290-20-2
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Solution
Strength 0.45 to 0.55 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR