Aminoguanidine hydrochloride 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

368910250 368911000
ACRO368910250EA 56.1 EUR
ACRO368910250 ACRO368911000
Aminoguanidine hydrochloride 98%
Aminoguanidine hydrochloride
Formule: CH₇ClN₄
Smeltpunt: >160 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00039074
CAS nummer: 1937-19-5
EINECS: 217-707-7

Order Now

Specification Test Results

Appearance White to off-white Crystals
Infrared spectrum Conforms
Melting point ≥160 °C
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR