4-Phenylbenzyl bromide 96%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 4-(Bromomethyl)biphenyl, 4-(Bromomethyl)-1,1'-biphenyl, p-(Bromomethyl)biphenyl

368950050 368950250
ACRO368950050EA 88.8 EUR
ACRO368950050 ACRO368950250
4-Phenylbenzyl bromide 96%
4-Phenylbenzyl bromide
Formule: C₁₃H₁₁Br
MW: 247.13 g/mol
Smeltpunt: 83…86 °C
Vlampunt: >110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00017869
CAS nummer: 2567-29-5
UN: 3261
ADR: 8 III

Order Now

Specification Test Results

Appearance Light yellow to beige Powder
Infrared spectrum Conforms
Melting point 83 to 86 °C
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR