Ethylenebis(triphenylphosphine)platinum(0) 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

369360010 369362500
ACRO369360010EA 382 EUR
ACRO369360010 ACRO369362500
Ethylenebis(triphenylphosphine)platinum(0) 98%
Ethylenebis(triphenylphosphine)platinum(0)
Formule: C₃₈H₃₄P₂Pt
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00012028
CAS nummer: 12120-15-9

Order Now

Specification Test Results

Appearance Light yellow to beige Powder
Infrared spectrum Conforms
Melting point 127 to 131 °C
Assay ≥97.5 % (ex Pt)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR