2-Bromo-5-chloropyridine 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

378980010 378980050
ACRO378980010EA 77.4 EUR
ACRO378980010 ACRO378980050
2-Bromo-5-chloropyridine 98%
2-Bromo-5-chloropyridine
Formule: C₅H₃BrClN
Smeltpunt: 65…69 °C
MDL Nummer: MFCD00234006
CAS nummer: 40473-01-6
UN: 2811

Order Now

Specification Test Results

Appearance Beige to yellow-brown Powder
Infrared spectrum Conforms
Melting point 65 to 69 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR