Tetraethylammonium perchlorate 0.2M in aqueous solution

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

379801000 379805000
ACRO379801000EA 40.6 EUR
ACRO379801000 ACRO379805000
Tetraethylammonium perchlorate 0.2M in aqueous solution
Tetraethylammonium perchlorate
Formule: C₈H₂₀ClNO₄
Smeltpunt: > 300 °C
Dichtheid: 1,0016 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00041977
CAS nummer: 2567-83-1
UN: 3139
ADR: 5.1,II

Order Now

Specification Test Results

Appearance Clear colorless Solution
Strength 0.2 M Exact strength on the certificate of analysis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR