4-Ethynylanisole 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

379920010 379920050
ACRO379920010EA 63.7 EUR
ACRO379920010 ACRO379920050
4-Ethynylanisole 97%
4-Ethynylanisole
Formule: C₉H₈O
Kookpunt: 87…91 °C (11 mmHg)
Smeltpunt: 28…30 °C
Dichtheid: 1,019 g/cm³
Vlampunt: 82 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00168815
CAS nummer: 768-60-5

Order Now

Specification Test Results

Appearance Clear colorless to yellow to brown Liquid or low melting solid
Infrared spectrum Conforms
Refractive index 1.5625 to 1.5675 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR