Calcium hydroxide 99.995% (trace metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

385610050 385610250
ACRO385610050EA 64.4 EUR
ACRO385610050 ACRO385610250
Calcium hydroxide 99.995% (trace metals basis)
Calcium hydroxide
Formule: Ca(OH)₂
Kookpunt: 2850 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010901
CAS nummer: 1305-62-0

Order Now

Specification Test Results

Appearance White Powder or crystals
Titration Complexometric ≥54 % (Ca)
Total trace metal impurities ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR