Glycine 99% ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Aminoacetic acid, Glycocoll, H-Gly-OH

385902500 385900500
ACRO385902500EA 56 EUR
ACRO385902500 ACRO385900500
Glycine 99% ACS
Glycine
Formule: NH₂CH₂COOH
Kookpunt: 181 °C (1013 hPa)
Smeltpunt: 240 °C
Dichtheid: 1,595 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008131
CAS nummer: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Sulfated ash ≤0.1 %
Titration with HClO4 ≥98.5 %
Substance darkened by H2SO4 Passes test
Chloride (Cl) ≤0.005 %
Sulfate (SO4) ≤0.005 %
Ammonium (NH4) ≤0.005 %
Heavy metals (as Pb) ≤0.002 %
Hydrolyzable substance Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR