Sodium cyclamate 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

389280010 389280500 389282500
ACRO389280010EA 128 EUR
ACRO389280010 ACRO389280500 ACRO389282500
Sodium cyclamate 99%
Sodium cyclamate
Formule: C₆H₁₂NNaO₃S
Smeltpunt: 265 °C
Vlampunt: >200 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003827
CAS nummer: 139-05-9

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Loss on drying ≤1 %
pH 5.5 to 7.5 (10 % solution)
Titration with HClO4 ≥98.5 % (On dry substance)
Sulfate (SO4) ≤0.1 %
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR