Ammoniak-oplossing 20% in water for analysis

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

390070010 390070025
ACRO390070010EA 27.4 EUR
ACRO390070010 ACRO390070025
Ammoniak-oplossing 20% in water for analysis
Ammoniak-oplossing
Formule: NH₃
Dichtheid: 0,92 g/cm³
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00011418
CAS nummer: 1336-21-6
UN: 2672
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear liquid
Color scale ≤10 APHA
Non-volatile matter ≤0.002 %
Titration with HCl ≥20 %
Chloride (Cl) ≤0.0001 %
Sulfide (S) ≤0.00001 %
Phosphate (PO4) ≤0.0001 %
Carbonate (as CO2) ≤0.002 %
Aluminium (Al) ≤0.00001 %
Antimony (Sb) ≤0.000002 %
Arsenic (As) ≤0.00001 %
Barium (Ba) ≤0.00001 %
Beryllium (Be) ≤0.00001 %
Bismuth (Bi) ≤0.000005 %
Cadmium (Cd) ≤0.00001 %
Calcium (Ca) ≤0.0001 %
Chromium (Cr) ≤0.000005 %
Cobalt (Co) ≤0.00001 %
Gallium (Ga) ≤0.000005 %
Germanium (Ge) ≤0.000002 %
Gold (Au) ≤0.00001 %
Indium (In) ≤0.000005 %
Iron (Fe) ≤0.00001 %
Lead (Pb) ≤0.000005 %
Lithium (Li) ≤0.000002 %
Magnesium (Mg) ≤0.00005 %
Manganese (Mn) ≤0.000001 %
Mercury (Hg) ≤0.00001 %
Molybdenum (Mo) ≤0.000002 %
Nickel (Ni) ≤0.000005 %
Platinum (Pt) ≤0.00001 %
Potassium (K) ≤0.0001 %
Silicon (Si) ≤0.00002 %
Silver (Ag) ≤0.000002 %
Sodium (Na) ≤0.0002 %
Strontium (Sr) ≤0.0001 %
Thallium (Tl) ≤0.000002 %
Tin (Sn) ≤0.000005 %
Titanium (Ti) ≤0.000005 %
Vanadium (V) ≤0.000002 %
Zinc (Zn) ≤0.00001 %
Zirconium (Zr) ≤0.000005 %
Heavy metals (as Pb) ≤1 ppm
KMnO4-reducing substance ≤0.001 % (as O)

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR