Ammoniumcarbonaat, extra pure

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Salt of hartshorn

390290010 390290025
ACRO390290010EA 43.5 EUR
ACRO390290010 ACRO390290025
Ammoniumcarbonaat, extra pure
Ammonium carbonate
Formule: (NH₄)₂CO₃
Dichtheid: 1,5 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010890
CAS nummer: 506-87-6
EINECS: 208-058-0
Merck Index: 13,00508

Order Now

Specification Test Results

Appearance White Powder or crystals and/or chunks
Non-volatile matter ≤100 ppm
Titration with HCl ≥30.5 % (NH3)
Chloride (Cl) ≤30 ppm
Sulfate (SO4) ≤30 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR