Calcium perchlorate tetrahydrate, pure

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

390631000 390635000
ACRO390631000EA 47.4 EUR
ACRO390631000 ACRO390635000
Calcium perchlorate tetrahydrate, pure
Calcium perchlorate tetrahydrate
Formule: CaClO₄·4H₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00191895
CAS nummer: 15627-86-8
UN: 1455
ADR: 5.1,II

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Titration Complexometric 12.2 to 13.5 % (Calcium)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR