N-Methyl-2-pyrrolidon (NMP) 99+% ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, 1-Methyl-2-pyrrolidone, NMP, Methyl Pyrrolidone

390682500 390680010 390680025 390680100
ACRO390682500EA 51.5 EUR
ACRO390682500 ACRO390680010 ACRO390680025 ACRO390680100
N-Methyl-2-pyrrolidon (NMP) 99+% ACS
N-Methyl-2-pyrrolidon (NMP)
Formule: C₅H₉NO
Kookpunt: 202 °C (760 mmHg)
Smeltpunt: –24 °C
Dichtheid: 1,03 g/cm³
Vlampunt: 91 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003193
CAS nummer: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.05 %
GC ≥99.50 %
Color scale ≤50 APHA
Chloride (Cl) ≤1 ppm
Aluminium (Al) ≤0.5 ppm
Barium (Ba) ≤0.1 ppm
Calcium (Ca) ≤0.5 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Nickel (Ni) ≤0.02 ppm
Zinc (Zn) ≤0.1 ppm
Free amine ≤0.01 % (as CH3NH2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR