Kaliumfluoride 40% (w/w) on alumina

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

390810250 390811000
ACRO390810250EA 86.4 EUR
ACRO390810250 ACRO390811000
Kaliumfluoride 40% (w/w) on alumina
Kaliumfluoride
Formule: KF
Kookpunt: 1505 °C (1013 hPa)
Smeltpunt: 860 °C
Dichtheid: 2,48 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011398
CAS nummer: 7789-23-3
UN: 1812
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White Powder
Assay 35 to 45 wt % (Mass balance)
Water 3 to 10 % (K.F.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR