Diammonium hydrogen citrate 98%, pure

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Citric acid diammonium salt, Diammonium hydrogen 2-hydroxypropane-1,2,3-tricarboxylate, Ammonium citrate dibasic

392505000 392500025 392501000
ACRO392505000EA 53 EUR
ACRO392505000 ACRO392500025 ACRO392501000
Diammonium hydrogen citrate 98%, pure
Diammonium hydrogen citrate
Formule: HOC(CO₂H)(CH₂CO₂NH₄)₂
Dichtheid: 1,5 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013068
CAS nummer: 3012-65-5

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Titration with HClO4 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR