Tetramethylammonium triacetoxyborohydride 90+%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

392850100 392850500
ACRO392850100EA 158 EUR
ACRO392850100 ACRO392850500
Tetramethylammonium triacetoxyborohydride 90+%
Tetramethylammonium triacetoxyborohydride
Formule: C₁₀H₂₂BNO₆
Smeltpunt: 95…98 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012196
CAS nummer: 109704-53-2
UN: 2813
ADR: 4.3,I

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Titration with HClO4 ≥90.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR