4-(Hydroxymethyl)benzoic acid 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

394340010 394340100
ACRO394340010EA 53.4 EUR
ACRO394340010 ACRO394340100
4-(Hydroxymethyl)benzoic acid 98%
4-(Hydroxymethyl)benzoic acid
Formule: C₈H₈O₃
Smeltpunt: 182…184 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00017598
CAS nummer: 3006-96-0

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 181 to 186 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR