4-(Trifluoromethyl)benzenesulphonyl chloride 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 4-(Trifluoromethyl)benzene-1-sulphonyl chloride

397760010 397760050
ACRO397760010EA 26.9 EUR
ACRO397760010 ACRO397760050
4-(Trifluoromethyl)benzenesulphonyl chloride 98%
4-(Trifluoromethyl)benzenesulphonyl chloride
Formule: C₇H₄ClF₃O₂S
MW: 244.62 g/mol
Kookpunt: 118…120 °C (18 mmHg)
Smeltpunt: 30…34 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00042422
CAS nummer: 2991-42-6
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Appearance White to light yellow Low melting solid
Infrared spectrum Conforms
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR