Bromomethyl acetate 95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

399180010 399180100
ACRO399180010EA 98.8 EUR
ACRO399180010 ACRO399180100
Bromomethyl acetate 95%
Bromomethyl acetate
Formule: H₃CCOOCH₂Br
Kookpunt: 130…133 °C (750 mmHg)
Vlampunt: 57 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000170
CAS nummer: 590-97-6
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4470 to 1.4540 (20°C, 589 nm)
GC ≥94 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR