Azelaic acid 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Azelaic acid,, 1,7-Heptanedicarboxylic acid, 1,9-Nonanedioic acid

401520250 401521000 401525000
ACRO401520250EA 29.4 EUR
ACRO401520250 ACRO401521000 ACRO401525000
Azelaic acid 98%
Azelaic acid
Formule: C₉H₁₆O₄
MW: 188.22 g/mol
Kookpunt: 237 °C (20hPa)
Smeltpunt: 98…103 °C
Vlampunt: 215 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004432
CAS nummer: 123-99-9
EINECS: 204-669-1
Merck Index: 13,00908

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR