(S)-(-)-4-Benzyl-2-oxazolidinone 99%, >99% ee

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

402160025 402160250
ACRO402160025EA 60.1 EUR
ACRO402160025 ACRO402160250
(S)-(-)-4-Benzyl-2-oxazolidinone 99%, >99% ee
(S)-(-)-4-Benzyl-2-oxazolidinone
Formule: C₁₀H₁₁NO₂
Smeltpunt: 87…90 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064496
CAS nummer: 90719-32-7

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 87 to 90 °C
HPLC ≥98.5 %
Enantiomeric excess ≥99 %
Specific optical rotation -60° to -64° (20 °C, 589 nm) (c=1, CHCl3)

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR