Bis(2-ethylhexyl)phosphate 95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Bis(2-ethylhexyl) hydrogen phosphate, Di-(2-ethylhexyl)phosphoric acid

402470010 402470050
ACRO402470010EA 219 EUR
ACRO402470010 ACRO402470050
Bis(2-ethylhexyl)phosphate 95%
Bis(2-ethylhexyl)phosphate
Formule: C₁₆H₃₅O₄P
Kookpunt: 48 °C (12 mmHg)
Smeltpunt: –60 °C
Dichtheid: 0,97 g/cm³
Vlampunt: 137 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009492
CAS nummer: 298-07-7
EINECS: 206-056-4
UN: 1902
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4400 to 1.4440 (20°C, 589 nm)
Titration with NaOH ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR