Sodium cholate 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

405191000 405195000
ACRO405191000EA 128 EUR
ACRO405191000 ACRO405195000
Sodium cholate 99%
Sodium cholate
Formule: C₂₄H₃₉NaO₅
Smeltpunt: 198 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00150749
CAS nummer: 361-09-1
EINECS: 206-643-5

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤5.0 % (110°C)
pH 7.5 to 9.5 (5 % in water)
Specific optical rotation +33° to +37° (20°C, 589 nm) (c=0.6, ethanol)
Titration with HClO4 98.0 to 102.0 % (On dried basis)
Heavy metals (as Pb) ≤50 ppm
Impurity ≤2.0 % Sodium deoxycholate


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR