Decyltrimethylammonium bromide 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: (1-Decyl)trimethylammonium bromide, N,N,N-Trimethyldecan-1-aminium bromide

406230250 406231000
ACRO406230250EA 70.6 EUR
ACRO406230250 ACRO406231000
Decyltrimethylammonium bromide 99%
Decyltrimethylammonium bromide
Formule: C₁₃H₃₀BrN
MW: 280.30 g/mol
Smeltpunt: 243 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00041973
CAS nummer: 2082-84-0
EINECS: 218-219-7

Order Now

Specification Test Results

Appearance White to beige Powder and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR