tetra-Chloroauric(III) acid trihydrate ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Gold(III) chloride trihydrate, Gold Chloride hydrate, Hydrogen tetrachloroaurate(III) trihydrate, Hydrogen tetrachloroaurate (III) trihydrate, tetra-Chloroauric acid trihydrate

411070010 411070050 411070250
ACRO411070010EA 183 EUR
ACRO411070010 ACRO411070050 ACRO411070250
tetra-Chloroauric(III) acid trihydrate ACS
tetra-Chloroauric(III) acid trihydrate
Formule: HAuCl₄·3H₂O
MW: 393.83 g/mol
Smeltpunt: 30
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00149904
CAS nummer: 16961-25-4
EINECS: 240-948-4
UN: 3260
ADR: 8 II

Order Now

Specification Test Results

Appearance Yellow to orange Powder and/or chunks (May have a brown hue)
Assay ≥49.0 % (Au)
Insoluble matter ≤0.1 % (10 % in ether)
Alkalies+oth.metals (SO4) ≤0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR