Hydroxylammonium chloride ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

412051000 412055000 412050025
ACRO412051000EA 45.7 EUR
ACRO412051000 ACRO412055000 ACRO412050025
Hydroxylammonium chloride ACS
Hydroxylammonium chloride
Formule: NH₂OH·HCl
Kookpunt: 306 °C (1013 hPa)
Smeltpunt: 155…158 °C
Dichtheid: 1,67 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00051089
CAS nummer: 5470-11-1
EINECS: 226-798-2
UN: 2923
ADR: 8,III
Merck Index: 13,04853

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Residue after ignition ≤0.05 %
Titration with KMnO4 ≥96.0 %
Clarity of alcohol solution Passes test
Ammonium (NH4) Passes test
Iron (Fe) ≤5 ppm
Heavy metals (ICP-OES) ≤5 ppm
Sulfur compounds (as SO4) ≤0.005 %
Titrable free acid ≤0.25 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR