Indigo carmine, pure, certified

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

412300250 412301000
ACRO412300250EA 47.7 EUR
ACRO412300250 ACRO412301000
Indigo carmine, pure, certified
Indigo carmine
Formule: C₁₆H₈N₂Na₂O₈S₂
Smeltpunt: ? 300 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005723
CAS nummer: 860-22-0
EINECS: 212-728-8
Merck Index: 13,04969

Order Now

Specification Test Results

Appearance Dark blue to purple Crystalline powder
Infrared spectrum Conforms
Dye content ≥80 %
UV Lambda max 608 to 612 nm
Absorption Ratio P-15/P+15 0.98 to 1.07
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR