Petroleum spirit, 35…60 °C, extra pure

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Gasoline, Petroleum ether, Petrol, Benzine

413140010
ACRO413140010EA 24.5 EUR
ACRO413140010
Petroleum spirit, 35…60 °C, extra pure
Petroleum spirit
Kookpunt: 35…60 °C (1013 hPa)
Vlampunt: –30 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00081849
CAS nummer: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3660 to 1.3710 (20°C, 589 nm)
Boiling point 35 to 60°C
Density (15°C) 0.649 to 0.659 g/ml
Color scale ≤10 APHA
Residue after evaporation ≤0.005 %
n-Hexane ≤3 %
Benzene ≤55 ppm
Aromatics ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR