Magnesium 99+%, ribbon

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

413380250 413381000
ACRO413380250EA 54.1 EUR
ACRO413380250 ACRO413381000
Magnesium 99+%, ribbon
Magnesium
Formule: Mg
Kookpunt: 1107 °C (1013 hPa)
Smeltpunt: 651 °C
Dichtheid: 1,74 g/cm³ (20 °C)
Vlampunt: 500 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00085308
CAS nummer: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1,III
Merck Index: 13,05675

Order Now

Specification Test Results

Appearance Silver white Metallic ribbon
Assay ≥99.0 %
Copper (Cu) ≤50 ppm
Iron (Fe) ≤500 ppm
Lead (Pb) ≤50 ppm
Nickel (Ni) ≤50 ppm
Zinc (Zn) ≤200 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR