Methylene blue hydrate, pure

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

414251000 414255000 414250025
ACRO414251000EA 59.8 EUR
ACRO414251000 ACRO414255000 ACRO414250025
Methylene blue hydrate, pure
Methylene blue hydrate
Formule: C₁₆H₁₈ClN₃S·xH₂O
Smeltpunt: 100…110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00150006
CAS nummer: 122965-43-9
EINECS: 200-515-2

Order Now

Specification Test Results

Appearance Green Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤22 %
Titration with TiCl3 ≥70 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR