Methyl-(±)-lactaat 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

414480250 414480050
ACRO414480250EA 159 EUR
ACRO414480250 ACRO414480050
Methyl-(±)-lactaat 99%
Methyl-(±)-lactaat
Formule: C₄H₈O₃
Kookpunt: 35 °C (6 mmHg)
Dichtheid: 1,09 g/cm³
Vlampunt: 49 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00066367
CAS nummer: 547-64-8
EINECS: 208-930-0
UN: 3272
ADR: 3,III
Merck Index: 12,06167

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4120 to 1.4150 (20°C, 589 nm)
GC ≥98.5 %
Specific optical rotation -0.5° to +0.5° (20°C, 589 nm) (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR