3-Nitrobenzenesulphonyl chloride 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

415840250 415841000
ACRO415840250EA 72.8 EUR
ACRO415840250 ACRO415841000
3-Nitrobenzenesulphonyl chloride 97%
3-Nitrobenzenesulphonyl chloride
Formule: C₆H₄ClNO₄S
Smeltpunt: 60…65 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007435
CAS nummer: 121-51-7
EINECS: 204-476-2
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance Yellow to orange light beige Crystalline powder
Infrared spectrum Conforms
Melting point 57 to 64 °C
HPLC ≥96.0 %
Specific optical rotation -105° to -109° (20 °C, 589 nm) (C=1, methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR