Fenolrood natriumzout ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

417250100 417250050 417251000
ACRO417250100EA 47 EUR
ACRO417250100 ACRO417250050 ACRO417251000
Fenolrood natriumzout ACS
Fenolrood natriumzout
Formule: C₁₉H₁₃NaO₅S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00066901
CAS nummer: 34487-61-1
EINECS: 252-057-8
Merck Index: 12,07397

Order Now

Specification Test Results

Appearance Dark red to red-brown Powder
Infrared spectrum Conforms
Visual transition interval From pH 6.8 (yellow) to pH 8.2 (red)
Clarity of solution Passes test

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR