Silver diethyldithiocarbamate ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: N,N-Diethyldithiocarbamic acid silver salt, Diethyldithiocarbamic acid silver salt, DETC, (Diethyldithiocarbamato-S,S')silver

419330100 419330250 419331000
ACRO419330100EA 101 EUR
ACRO419330100 ACRO419330250 ACRO419331000
Silver diethyldithiocarbamate ACS
Silver diethyldithiocarbamate
Formule: (C₂H₅)₂NCSSAg
MW: 256.14 g/mol
Smeltpunt: 172…175 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00004929
CAS nummer: 1470-61-7
EINECS: 216-003-7

Order Now

Specification Test Results

Appearance Green to khaki Powder
Infrared spectrum Conforms
Solubility (in Pyridine) Passes test
Suitability (absorbance < 0.1) Passes test (for As determination)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR