Sodium carbonate monohydrate 99.6% ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Sodium carbonate hydrous, Soda monohydrate

419480010 419480051
ACRO419480010EA 45.6 EUR
ACRO419480010 ACRO419480051
Sodium carbonate monohydrate 99.6% ACS
Sodium carbonate monohydrate
Formule: Na₂CO₃·1H₂O
MW: 124.00 g/mol
Smeltpunt: 851 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149177
CAS nummer: 5968-11-6
EINECS: 207-838-8

Order Now

Specification Test Results

Appearance White Crystalline powder
Loss on drying 13.0 % to 15.0 % (150°C)
Titration with HCl ≥99.5 %
Chloride (Cl) ≤10 ppm
Phosphate (PO4) ≤5 ppm
Calcium (Ca) ≤300 ppm
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤50 ppm
Potassium (K) ≤50 ppm
Heavy metals (as Pb) ≤5 ppm
Silicon dioxide (SiO2) ≤50 ppm
Sulfur compounds (as SO4) ≤0.004 %
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR