Tris(2-methylphenyl)phosphine 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

422320050 422320010 422320250
ACRO422320050EA 161 EUR
ACRO422320050 ACRO422320010 ACRO422320250
Tris(2-methylphenyl)phosphine 99%
Tris(2-methylphenyl)phosphine
Formule: C₂₁H₂₁P
Kookpunt: 187 - 192 °C (0,05 -rr)
Smeltpunt: 126…127 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008514
CAS nummer: 6163-58-2
EINECS: 228-193-9

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 122 to 126 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR